Learn More
Kyfora Bio DSPE PEG 2000 Amine 10 MG
Catalog No. 502559797
Encompass_Preferred
DSPE-PEG(2000)-amine is DSPE (a phospholipid) functionalized with PEG2000. It can be used in the synthesis of solid lipid and thermosensitive nanoparticles, as well as for encapsulation of hydrophobic drugs. Functionalization with PEG prevents increases bioavailability and prevents opsonization. It has been used for the delivery of anticancer agents as well as other drug cargo. Key application(s): Drug delivery
Catalog No. 50-255-9797 Supplier Kyfora Bio Supplier No. 1449910
DSPE-PEG(2000)-amine is DSPE (a phospholipid) functionalized with PEG2000. It can be used in the synthesis of solid lipid and thermosensitive nanoparticles, as well as for encapsulation of hydrophobic drugs. Functionalization with PEG prevents increases bioavailability and prevents opsonization. It has been used for the delivery of anticancer agents as well as other drug cargo. Key application(s): Drug delivery