Learn More
Medchemexpress LLC DSPE-PEG(2000) amine (ammonium salt) | 474922-26-4 | ≥95.0% | 2790.52 g/mol | C46H94N3O11P | 100MG
SDP
Catalog No. 5000236376
Encompass_Preferred
DSPE-PEG(2000) amine (ammonium salt) is an amine-terminated, PEGylated phospholipid used to introduce reactive primary amine groups onto the surface of liposomes and lipid nanoparticles for covalent conjugation. The hydrophobic DSPE tail anchors the molecule in lipid bilayers while the PEG 2000 spacer provides steric stabilization and improves aqueous solubility; the terminal amine enables coupling to activated esters, dyes, linkers, or targeting ligands.
Catalog No. 50-002-36376 Supplier Medchemexpress LLC Supplier No. HY125924100MG
DSPE-PEG(2000) amine (ammonium salt) is an amine-terminated, PEGylated phospholipid used to introduce reactive primary amine groups onto the surface of liposomes and lipid nanoparticles for covalent conjugation. The hydrophobic DSPE tail anchors the molecule in lipid bilayers while the PEG 2000 spacer provides steric stabilization and improves aqueous solubility; the terminal amine enables coupling to activated esters, dyes, linkers, or targeting ligands.