DSPE-PEG(2000) Maleimide is a highly purified, functionalized polyethylene glycol (PEG) lipid designed for advanced research applications in targeted drug delivery. This lipid enables precise conjugation of bioactive molecules to nanoparticle systems, ensuring reliable results in therapies for genetic disorders, cancer, and neurological conditions. Its exceptional purity is confirmed through HNMR analysis, making it a preferred choice for researchers focused on enhancing the specificity and efficacy of drug delivery systems.
- Highly purified PEG lipid for advanced drug delivery
- Facilitates conjugation with thiol-containing bioactive molecules
- Verified structural integrity with no detectable maleimide ring opening
- Application in targeted therapies for genetic disorders and cancer
- Stored at -20°C and shipped with dry ice for optimal stability
- Purity greater than 99% verified by TLC