Learn More
Avanti Polar Lipids DSPE-PEG(2000)MALEIMIDE 50MG
Catalog No. NC9159499
Encompass
DSPE-PEG(2000)Maleimide, Molecular Weight: 2941.64, Molecular Formula: C 139 H 271 N4O 57 P, Powder form, White Color, Not Flammable, 50mg
Catalog No. NC9159499 Supplier Avanti Polar Lipids Supplier No. 880126P50MG
DSPE-PEG(2000)Maleimide, Molecular Weight: 2941.64, Molecular Formula: C 139 H 271 N4O 57 P, Powder form, White Color, Not Flammable, 50mg