Learn More
Medchemexpress LLC Dspe-peg-alkyne | 85.0% | 2000 | 25 MG
SDP
Catalog No. 5000395183
Encompass_Preferred
DSPE-PEG-alkyne (MW 2000) is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. It can be used in drug delivery research.
  • Peg lipid
  • Improves delivery efficiency
  • Enhances tissue specificity for poorly soluble drugs
  • Suitable for drug delivery research
Catalog No. 50-003-95183 Supplier Medchemexpress LLC Supplier No. 25MGHY167028
DSPE-PEG-alkyne (MW 2000) is a PEG lipid that can improve the delivery efficiency and tissue specificity of poorly soluble drugs. It can be used in drug delivery research.
  • Peg lipid
  • Improves delivery efficiency
  • Enhances tissue specificity for poorly soluble drugs
  • Suitable for drug delivery research