Learn More
Medchemexpress LLC DSPE-PEG-Amine | 95.0% | 500 MG
SDP
Catalog No. 5000354943
Encompass_Preferred
DSPE-PEG-Amine, MW 5000 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligand for an E3 ubiquitin ligase
  • Ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54943 Supplier Medchemexpress LLC Supplier No. HY140735500MG
DSPE-PEG-Amine, MW 5000 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligand for an E3 ubiquitin ligase
  • Ligand for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins