Learn More
Medchemexpress LLC Dspe-peg-amine, ammonium | 474922-26-4 | 100.0% | 2000 (Average) | (C2H4O)nC44H87N2O10P.NH3 | 50MG
SDP
Catalog No. 5000236374
Encompass_Preferred
DSPE-PEG-Amine, MW 2000 (ammonium) is an amine-terminated polyethylene glycol-conjugated phospholipid used to introduce reactive primary amine groups onto the surface of liposomes and lipid nanoparticles. It is commonly used for bioconjugation, coupling chemistries, and preparing functionalized delivery systems in drug delivery research.
  • Amine-terminated PEG-lipid provides a reactive primary amine for coupling chemistry.
  • Average molecular weight of 2000 supports colloidal stability and stealth properties.
  • High purity suitable for research and formulation development.
  • Solid form with recommended storage at -20°C; protect from light.
  • Small pack sizes facilitate screening and optimization of formulations.
  • Compatible with NHS-ester and activated carboxyl conjugation methods.
Catalog No. 50-002-36374 Supplier Medchemexpress LLC Supplier No. HY12592450MG
DSPE-PEG-Amine, MW 2000 (ammonium) is an amine-terminated polyethylene glycol-conjugated phospholipid used to introduce reactive primary amine groups onto the surface of liposomes and lipid nanoparticles. It is commonly used for bioconjugation, coupling chemistries, and preparing functionalized delivery systems in drug delivery research.
  • Amine-terminated PEG-lipid provides a reactive primary amine for coupling chemistry.
  • Average molecular weight of 2000 supports colloidal stability and stealth properties.
  • High purity suitable for research and formulation development.
  • Solid form with recommended storage at -20°C; protect from light.
  • Small pack sizes facilitate screening and optimization of formulations.
  • Compatible with NHS-ester and activated carboxyl conjugation methods.