Learn More
Medchemexpress LLC Dspe-peg-amine (ammonium) | 474922-26-4 | 98.8% | 1000 (average) | (C2H4O)nN2O10P·NH3 | 25 MG
SDP
Catalog No. 5000281191
Encompass_Preferred
DSPE-PEG-Amine (average MW 1000, ammonium salt) is a PEGylated phospholipid with a terminal primary amine used to functionalize liposomes and nanoparticles. The hydrophobic DSPE anchor incorporates into lipid bilayers while the PEG spacer presents a reactive amine at the particle surface for covalent conjugation to ligands, dyes, or biomolecules in delivery and surface-modification applications.
  • PEGylated phospholipid with terminal primary amine.
  • Average molecular weight approximately 1000 daltons.
  • Ammonium salt form to aid handling and solubility.
  • Enables covalent conjugation to NHS-esters, aldehydes, and other activated groups.
  • Integrates into liposome bilayers for surface functionalization.
  • Suitable for nanoparticle coating and targeted delivery research.
Catalog No. 50-002-81191 Supplier Medchemexpress LLC Supplier No. HYW440823A25MG
DSPE-PEG-Amine (average MW 1000, ammonium salt) is a PEGylated phospholipid with a terminal primary amine used to functionalize liposomes and nanoparticles. The hydrophobic DSPE anchor incorporates into lipid bilayers while the PEG spacer presents a reactive amine at the particle surface for covalent conjugation to ligands, dyes, or biomolecules in delivery and surface-modification applications.
  • PEGylated phospholipid with terminal primary amine.
  • Average molecular weight approximately 1000 daltons.
  • Ammonium salt form to aid handling and solubility.
  • Enables covalent conjugation to NHS-esters, aldehydes, and other activated groups.
  • Integrates into liposome bilayers for surface functionalization.
  • Suitable for nanoparticle coating and targeted delivery research.