Learn More
Medchemexpress LLC DSPE-PEG-Amine, MW 5000 | 95.0% | 100 MG
SDP
Catalog No. 5000356654
Encompass_Preferred
DSPE-PEG-Amine, MW 5000 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound is intended for research use only. PROTACs function by connecting two ligands, one for an E3 ubiquitin ligase and another for a target protein, to exploit the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
  • Average molecular weight of 5000
  • Appears as a solid, white to off-white material
Catalog No. 50-003-56654 Supplier Medchemexpress LLC Supplier No. HY140735100MG
DSPE-PEG-Amine, MW 5000 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound is intended for research use only. PROTACs function by connecting two ligands, one for an E3 ubiquitin ligase and another for a target protein, to exploit the intracellular ubiquitin-proteasome system for selective protein degradation.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed for selective degradation of target proteins
  • Average molecular weight of 5000
  • Appears as a solid, white to off-white material