Learn More
Medchemexpress LLC DSPE-PEG-Amine, MW 5000 | 95.0% | 50 MG
SDP
Catalog No. 5000353521
Encompass_Preferred
DSPE-PEG-Amine, MW 5000 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules connect two ligands, one for an E3 ubiquitin ligase and another for a target protein, leveraging the intracellular ubiquitin-proteasome system to selectively degrade specific proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53521 Supplier Medchemexpress LLC Supplier No. HY14073550MG
DSPE-PEG-Amine, MW 5000 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These molecules connect two ligands, one for an E3 ubiquitin ligase and another for a target protein, leveraging the intracellular ubiquitin-proteasome system to selectively degrade specific proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins