Learn More
Medchemexpress LLC DSPE-PEG-azide | 90.6% | 2000 (average) | C44H85N4O10P | 10 MG
SDP
Catalog No. 5000281271
Encompass_Preferred
DSPE-PEG-azide (average MW 2000) is an azide-functionalized phospholipid-polyethylene glycol conjugate used to introduce reactive azide handles on lipid assemblies. It enables click-chemistry bioconjugation for surface modification of liposomes, micelles, and nanoparticles in drug-delivery and biofunctionalization workflows.
  • Provides an azide functional group compatible with CuAAC and SPAAC reactions.
  • Facilitates stable PEGylation for improved colloidal stability and circulation.
  • Suitable for incorporation into liposomes, micelles, and other lipid nanoparticles.
  • Supplied as a small-quantity reagent for research-scale conjugation workflows.
  • Characterized by an average molecular weight of 2000 and a purity of 90.6%.
Catalog No. 50-002-81271 Supplier Medchemexpress LLC Supplier No. HYW44083210MG
DSPE-PEG-azide (average MW 2000) is an azide-functionalized phospholipid-polyethylene glycol conjugate used to introduce reactive azide handles on lipid assemblies. It enables click-chemistry bioconjugation for surface modification of liposomes, micelles, and nanoparticles in drug-delivery and biofunctionalization workflows.
  • Provides an azide functional group compatible with CuAAC and SPAAC reactions.
  • Facilitates stable PEGylation for improved colloidal stability and circulation.
  • Suitable for incorporation into liposomes, micelles, and other lipid nanoparticles.
  • Supplied as a small-quantity reagent for research-scale conjugation workflows.
  • Characterized by an average molecular weight of 2000 and a purity of 90.6%.