Learn More
Medchemexpress LLC DSPE-PEG-azide | 90.6% | 2000 Da (average) | 25 MG
SDP
Catalog No. 5000281459
Encompass_Preferred
DSPE-PEG-Azide (MW 2000) is an azide-functionalized PEG-lipid supplied as a white to off-white solid for research use. It provides an azide handle for click-chemistry conjugation and is used to PEGylate lipid surfaces to modify nanoparticle and liposome properties.
  • Provides azide functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Enables surface PEGylation of liposomes and nanoparticles to improve colloidal stability and circulation.
  • Average molecular weight about 2000 Da, giving defined PEG chain length.
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo dosing at ≥2.5 mg/mL with common vehicles.
  • Supplied as a white to off-white solid; store powder at -20 °C.
  • Reported purity approximately 90.6%.
Catalog No. 50-002-81459 Supplier Medchemexpress LLC Supplier No. HYW44083225MG
DSPE-PEG-Azide (MW 2000) is an azide-functionalized PEG-lipid supplied as a white to off-white solid for research use. It provides an azide handle for click-chemistry conjugation and is used to PEGylate lipid surfaces to modify nanoparticle and liposome properties.
  • Provides azide functional group for copper-catalyzed azide-alkyne cycloaddition.
  • Enables surface PEGylation of liposomes and nanoparticles to improve colloidal stability and circulation.
  • Average molecular weight about 2000 Da, giving defined PEG chain length.
  • Soluble in DMSO (100 mg/mL) and formulatable for in vivo dosing at ≥2.5 mg/mL with common vehicles.
  • Supplied as a white to off-white solid; store powder at -20 °C.
  • Reported purity approximately 90.6%.