Learn More
Medchemexpress LLC Dspe-peg-biotin | 385437-57-0 | 99.9% | 50 MG
SDP
Catalog No. 5000352965
Encompass_Preferred
Dspe-peg-biotin is a Peg-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand is tailored to bind to an E3 ubiquitin ligase, while the other targets a specific protein. This mechanism enables PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • PROTACs are composed of two distinct ligands joined by a linker.
  • One ligand specifically targets an E3 ubiquitin ligase.
  • The other ligand binds to the protein intended for degradation.
  • Operates by harnessing the intracellular ubiquitin-proteasome system.
  • Facilitates the selective degradation of target proteins.
Catalog No. 50-003-52965 Supplier Medchemexpress LLC Supplier No. HY14073650MG
Dspe-peg-biotin is a Peg-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand is tailored to bind to an E3 ubiquitin ligase, while the other targets a specific protein. This mechanism enables PROTACs to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based PROTAC linker.
  • Utilized in the synthesis of PROTACs.
  • PROTACs are composed of two distinct ligands joined by a linker.
  • One ligand specifically targets an E3 ubiquitin ligase.
  • The other ligand binds to the protein intended for degradation.
  • Operates by harnessing the intracellular ubiquitin-proteasome system.
  • Facilitates the selective degradation of target proteins.