Learn More
Medchemexpress LLC DSPE-PEG-biotin | 385437-57-0 | 99.9% | C142H280N5O56PS | 10MG
SDP
Catalog No. 5000258806
Encompass_Preferred
DSPE-PEG-Biotin (average molecular weight 2000) is an amphiphilic lipid-polyethylene glycol conjugate bearing a terminal biotin. It enables incorporation of biotin functionality into lipid assemblies, liposomes, micelles, and biomolecules for affinity capture, surface functionalization, and use as a PEG-based linker in conjugation chemistries.
  • Amphiphilic lipid-PEG conjugate enabling membrane incorporation.
  • Terminal biotin enables strong binding to streptavidin and avidin.
  • Average molecular weight 2000 and high purity suitable for conjugation.
  • Soluble in DMSO (25 mg/mL) and dispersible in water (5 mg/mL) with ultrasonic aid.
  • Stable as a powder at -20°C; solvent stability is limited.
  • Useful for liposome functionalization, affinity capture, and linker synthesis in bioconjugation.
Catalog No. 50-002-58806 Supplier Medchemexpress LLC Supplier No. HY14073610MG
DSPE-PEG-Biotin (average molecular weight 2000) is an amphiphilic lipid-polyethylene glycol conjugate bearing a terminal biotin. It enables incorporation of biotin functionality into lipid assemblies, liposomes, micelles, and biomolecules for affinity capture, surface functionalization, and use as a PEG-based linker in conjugation chemistries.
  • Amphiphilic lipid-PEG conjugate enabling membrane incorporation.
  • Terminal biotin enables strong binding to streptavidin and avidin.
  • Average molecular weight 2000 and high purity suitable for conjugation.
  • Soluble in DMSO (25 mg/mL) and dispersible in water (5 mg/mL) with ultrasonic aid.
  • Stable as a powder at -20°C; solvent stability is limited.
  • Useful for liposome functionalization, affinity capture, and linker synthesis in bioconjugation.