Learn More
Medchemexpress LLC Dspe-peg-biotin | 385437-57-0 | 99.93% | 100 MG
SDP
Catalog No. 5000352964
Encompass_Preferred
DSPE-PEG-Biotin, MW 2000 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Can be dissolved in DMSO and H2O for in vitro experiments.
  • Multiple dissolution protocols are available for in vivo experiments using various co-solvents like DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.
Catalog No. 50-003-52964 Supplier Medchemexpress LLC Supplier No. HY140736100MG
DSPE-PEG-Biotin, MW 2000 is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Can be dissolved in DMSO and H2O for in vitro experiments.
  • Multiple dissolution protocols are available for in vivo experiments using various co-solvents like DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.