Learn More
Medchemexpress LLC DSPE-PEG-biotin | 98.4% | 3,400 (average) | 10 MG
SDP
Catalog No. 5000281643
Encompass_Preferred
DSPE-PEG-Biotin is an amphiphilic phospholipid-polyethylene glycol conjugate functionalized with biotin (average molecular weight 3,400). Supplied as a white solid, it is used to introduce biotin groups into lipid membranes and liposomal formulations for labeling, affinity capture, and targeted delivery studies in biochemical and drug-delivery research. Powder storage and solvent storage recommendations are provided by the manufacturer.
Catalog No. 50-002-81643 Supplier Medchemexpress LLC Supplier No. HYW44088610MG
DSPE-PEG-Biotin is an amphiphilic phospholipid-polyethylene glycol conjugate functionalized with biotin (average molecular weight 3,400). Supplied as a white solid, it is used to introduce biotin groups into lipid membranes and liposomal formulations for labeling, affinity capture, and targeted delivery studies in biochemical and drug-delivery research. Powder storage and solvent storage recommendations are provided by the manufacturer.