Learn More
Medchemexpress LLC Dspe-peg-biotin, MW 2000 | 385437-57-0 | 99.9% | 25 MG
SDP
Catalog No. 5000352680
Encompass_Preferred
DSPE-PEG-Biotin, MW 2000 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Facilitates PROTAC synthesis
  • Targets proteins for degradation
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-52680 Supplier Medchemexpress LLC Supplier No. 25MGHY140736
DSPE-PEG-Biotin, MW 2000 is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Facilitates PROTAC synthesis
  • Targets proteins for degradation
  • Utilizes the ubiquitin-proteasome system