Learn More
Medchemexpress LLC DSPE-PEG-biotin, MW 3400 | 98.4% | 3400 (average) | 250 MG
SDP
Catalog No. 5000281262
Encompass_Preferred
DSPE-PEG-Biotin, MW 3400 is a PEGylated phospholipid supplied as a white powder. The DSPE lipid tail anchors in membranes or liposomes while the PEG3400 spacer presents biotin for high-affinity capture, detection, or targeted delivery applications. The product is listed with an average molecular weight of 3400 and a reported purity of 98.43%.
  • Provides biotin functional groups for streptavidin/avidin-based capture and detection.
  • An amphiphilic structure that anchors efficiently into lipid bilayers and liposomes.
  • PEG3400 spacer improves aqueous solubility and reduces nonspecific interactions.
  • Available in multiple pack sizes for small- and preparative-scale work.
  • Stable as a powder at -20°C; in-solution storage recommendations provided for longer-term stability.
Catalog No. 50-002-81262 Supplier Medchemexpress LLC Supplier No. HYW440886250MG
DSPE-PEG-Biotin, MW 3400 is a PEGylated phospholipid supplied as a white powder. The DSPE lipid tail anchors in membranes or liposomes while the PEG3400 spacer presents biotin for high-affinity capture, detection, or targeted delivery applications. The product is listed with an average molecular weight of 3400 and a reported purity of 98.43%.
  • Provides biotin functional groups for streptavidin/avidin-based capture and detection.
  • An amphiphilic structure that anchors efficiently into lipid bilayers and liposomes.
  • PEG3400 spacer improves aqueous solubility and reduces nonspecific interactions.
  • Available in multiple pack sizes for small- and preparative-scale work.
  • Stable as a powder at -20°C; in-solution storage recommendations provided for longer-term stability.