Learn More
Medchemexpress LLC DSPE-PEG carboxylic acid (sodium) | 1403744-37-5 | 2000 (average) | (C2H4O)nC45H86NO12P.Na | 5 MG
SDP
Catalog No. 5000221542
Encompass_Preferred
DSPE-PEG carboxylic acid (sodium), MW 2000 is a polyethylene glycol-conjugated phospholipid with a terminal carboxylate (sodium salt). It is used as a PEG-lipid component for formulating liposomes and nanoparticle drug delivery systems to improve circulation time, stability, and surface functionalization.
  • PEG-lipid with terminal carboxylate (sodium salt) enabling conjugation and surface modification.
  • Average molecular weight approximately 2000, suitable for PEGylation and stealth coatings.
  • Improves liposome circulation time and colloidal stability.
  • Enhances targeting potential and cellular uptake when formulated into nanoparticles.
  • Solid, white to off-white form for convenient handling and storage.
Catalog No. 50-002-21542 Supplier Medchemexpress LLC Supplier No. HY141892A5MG
DSPE-PEG carboxylic acid (sodium), MW 2000 is a polyethylene glycol-conjugated phospholipid with a terminal carboxylate (sodium salt). It is used as a PEG-lipid component for formulating liposomes and nanoparticle drug delivery systems to improve circulation time, stability, and surface functionalization.
  • PEG-lipid with terminal carboxylate (sodium salt) enabling conjugation and surface modification.
  • Average molecular weight approximately 2000, suitable for PEGylation and stealth coatings.
  • Improves liposome circulation time and colloidal stability.
  • Enhances targeting potential and cellular uptake when formulated into nanoparticles.
  • Solid, white to off-white form for convenient handling and storage.