Learn More
Medchemexpress LLC DSPE-PEG carboxylic acid (sodium) | 1403744-37-5 | 98.0% | (C2H4O)nC45H86NO12P.Na | 25 MG
SDP
Catalog No. 5000352621
Encompass_Preferred
DSPE-PEG Carboxylic acid (sodium), MW 2000 is a PEG-lipid that can be used in the preparation of drug delivery nanoparticles. It is capable of extending the blood circulation time and stability of liposomes, while also improving the targeting ability and cellular uptake efficiency of nanoparticles.
  • Used in the preparation of drug delivery nanoparticles
  • Prolongs the blood circulation time and stability of liposomes
  • Enhances the targeting ability of nanoparticles
  • Improves cellular uptake efficiency of nanoparticles
Catalog No. 50-003-52621 Supplier Medchemexpress LLC Supplier No. 25MGHY141892A
DSPE-PEG Carboxylic acid (sodium), MW 2000 is a PEG-lipid that can be used in the preparation of drug delivery nanoparticles. It is capable of extending the blood circulation time and stability of liposomes, while also improving the targeting ability and cellular uptake efficiency of nanoparticles.
  • Used in the preparation of drug delivery nanoparticles
  • Prolongs the blood circulation time and stability of liposomes
  • Enhances the targeting ability of nanoparticles
  • Improves cellular uptake efficiency of nanoparticles