Learn More
Medchemexpress LLC Dspe-peg carboxylic acid (sodium), MW 2000 | 1403744-37-5 | 98.0% | (C2H4O)nC45H86NO12P.Na | 100 MG
SDP
Catalog No. 5000353608
Encompass_Preferred
DSPE-PEG Carboxylic acid (sodium), MW 2000 is a PEG-lipid used in the preparation of drug delivery nanoparticles. It can prolong the blood circulation time and stability of liposomes, and enhance the targeting ability and cellular uptake efficiency of nanoparticles. The molecular weight is 2000 (Average), and it appears as a white to off-white solid.
  • Used in preparation of drug delivery nanoparticles
  • Can prolong blood circulation time and stability of liposomes
  • Enhances targeting ability
  • Enhances cellular uptake efficiency of nanoparticles
  • For research use only
Catalog No. 50-003-53608 Supplier Medchemexpress LLC Supplier No. HY141892A100MG
DSPE-PEG Carboxylic acid (sodium), MW 2000 is a PEG-lipid used in the preparation of drug delivery nanoparticles. It can prolong the blood circulation time and stability of liposomes, and enhance the targeting ability and cellular uptake efficiency of nanoparticles. The molecular weight is 2000 (Average), and it appears as a white to off-white solid.
  • Used in preparation of drug delivery nanoparticles
  • Can prolong blood circulation time and stability of liposomes
  • Enhances targeting ability
  • Enhances cellular uptake efficiency of nanoparticles
  • For research use only