Learn More
Medchemexpress LLC Dspe-peg-cho | 89.0% | 2000 (Average) | 50 MG
SDP
Catalog No. 5000387439
Encompass_Preferred
DSPE-PEG-CHO (MW 2000) is a linear heterobifunctional PEGylation reagent featuring DSPE phospholipids and aldehyde groups. Its PEG linkers provide good hydrophilicity and water solubility. This reagent is commonly used to bind antibodies, peptides, or other ligands to the surface of liposomes and lipid-polyethylene glycol nanoparticles.
  • Linear heterobifunctional PEGylation reagent with DSPE phospholipids and aldehyde groups.
  • PEG linkers provide good hydrophilicity and water solubility.
  • Used to bind antibodies, peptides, or other ligands to liposomes and lipid-polyethylene glycol nanoparticles.
  • Can be used for drug delivery.
Catalog No. 50-003-87439 Supplier Medchemexpress LLC Supplier No. HY176509A50MG
DSPE-PEG-CHO (MW 2000) is a linear heterobifunctional PEGylation reagent featuring DSPE phospholipids and aldehyde groups. Its PEG linkers provide good hydrophilicity and water solubility. This reagent is commonly used to bind antibodies, peptides, or other ligands to the surface of liposomes and lipid-polyethylene glycol nanoparticles.
  • Linear heterobifunctional PEGylation reagent with DSPE phospholipids and aldehyde groups.
  • PEG linkers provide good hydrophilicity and water solubility.
  • Used to bind antibodies, peptides, or other ligands to liposomes and lipid-polyethylene glycol nanoparticles.
  • Can be used for drug delivery.