Learn More
Medchemexpress LLC DSPE-PEG-CHO (aldehyde-terminated DSPE-PEG) | 2000 (average) | 10 MG
SDP
Catalog No. 5000294709
Encompass_Preferred
DSPE-PEG-CHO (MW 2000) is a heterobifunctional PEGylated phospholipid composed of a DSPE lipid anchor linked to a polyethylene glycol chain terminated with an aldehyde group. Supplied as a powder, it is designed for introducing reactive aldehyde functionality onto liposomes, lipid nanoparticles, or other lipid-based carriers to enable conjugation of biomolecules for targeted delivery and surface modification.
  • Aldehyde-terminated PEG enables covalent conjugation to primary amines.
  • PEG MW 2000 provides hydrophilicity and steric stabilization of particles.
  • DSPE lipid anchor facilitates efficient incorporation into lipid bilayers and nanoparticles.
  • DMSO solubility approximately 12.5 mg/mL with ultrasonic warming to 60°C.
  • Powder storage at -20°C (up to 3 years); in solvent storage at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-002-94709 Supplier Medchemexpress LLC Supplier No. HY176509A10MG
DSPE-PEG-CHO (MW 2000) is a heterobifunctional PEGylated phospholipid composed of a DSPE lipid anchor linked to a polyethylene glycol chain terminated with an aldehyde group. Supplied as a powder, it is designed for introducing reactive aldehyde functionality onto liposomes, lipid nanoparticles, or other lipid-based carriers to enable conjugation of biomolecules for targeted delivery and surface modification.
  • Aldehyde-terminated PEG enables covalent conjugation to primary amines.
  • PEG MW 2000 provides hydrophilicity and steric stabilization of particles.
  • DSPE lipid anchor facilitates efficient incorporation into lipid bilayers and nanoparticles.
  • DMSO solubility approximately 12.5 mg/mL with ultrasonic warming to 60°C.
  • Powder storage at -20°C (up to 3 years); in solvent storage at -80°C for 6 months or -20°C for 1 month.