Learn More
Medchemexpress LLC DSPE-PEG-CHO (MW 2000) | 89.0% | 2000 | 25 MG
SDP
Catalog No. 5000384097
Encompass_Preferred
This product is DSPE-PEG-CHO with an average molecular weight of 2000. It is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Purity of 89.04% by HPLC
  • 1H NMR spectrum consistent with its structure
Catalog No. 50-003-84097 Supplier Medchemexpress LLC Supplier No. 25MGHY176509A
This product is DSPE-PEG-CHO with an average molecular weight of 2000. It is intended for research use only and has not been fully validated for medical applications.
  • White to off-white solid appearance
  • Purity of 89.04% by HPLC
  • 1H NMR spectrum consistent with its structure