Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-CHO (MW 2000) | 89.04% | 2000 (Average) | 100 MG
SDP

Catalog No. 5000387267
Encompass_Preferred

DSPE-PEG-CHO (MW 2000) is a high-purity chemical compound from MedChemExpress, identified by catalog number HY-176509A. This white to off-white solid has an average molecular weight of 2000 and a purity of 89.04% as determined by HPLC. It is suitable for a variety of research applications and requires specific storage conditions to maintain its stability.

  • White to off-white solid appearance
  • Purity of 89.04% (HPLC)
  • Average molecular weight of 2000
  • Recommended storage at -20°C for up to 3 years as powder
  • Recommended storage at -80°C for 6 months, or -20°C for 1 month when in solvent
  • For research use only

Catalog No. 50-003-87267 Supplier Medchemexpress LLC Supplier No. HY176509A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-CHO (MW 2000) is a high-purity chemical compound from MedChemExpress, identified by catalog number HY-176509A. This white to off-white solid has an average molecular weight of 2000 and a purity of 89.04% as determined by HPLC. It is suitable for a variety of research applications and requires specific storage conditions to maintain its stability.

  • White to off-white solid appearance
  • Purity of 89.04% (HPLC)
  • Average molecular weight of 2000
  • Recommended storage at -20°C for up to 3 years as powder
  • Recommended storage at -80°C for 6 months, or -20°C for 1 month when in solvent
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.