Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-COOH | 98.1% | 5000 | 100 MG
SDP

Catalog No. 5000390794
Encompass_Preferred

DSPE-PEG-COOH (MW 5000) is a PEG lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for various applications in drug delivery research.

  • Improves delivery of poorly soluble drugs.
  • Enhances tissue specificity.
  • Solid, white to off-white appearance.
  • Recommended storage at -20°C, sealed, away from moisture and light.
  • Soluble in DMSO at 100 mg/mL (with ultrasonic assistance).

Catalog No. 50-003-90794 Supplier Medchemexpress LLC Supplier No. HY166968100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-COOH (MW 5000) is a PEG lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for various applications in drug delivery research.

  • Improves delivery of poorly soluble drugs.
  • Enhances tissue specificity.
  • Solid, white to off-white appearance.
  • Recommended storage at -20°C, sealed, away from moisture and light.
  • Soluble in DMSO at 100 mg/mL (with ultrasonic assistance).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.