Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-COOH | 98.1% | 5000 (average) | 10 MG
SDP

Catalog No. 5000289315
Encompass_Preferred

DSPE-PEG-COOH (MW 5000) is a PEGylated phospholipid used to modify liposome and nanoparticle surfaces to improve solubility, circulation time, and tissue targeting of poorly soluble drugs.

  • PEGylated phospholipid with an average molecular weight of 5,000 Da.
  • Carboxylic acid terminal group enables covalent conjugation and surface functionalization.
  • Enhances colloidal stability and circulation half-life in delivery systems.
  • Soluble in DMSO at approximately 100 mg/mL with sonication; hygroscopic DMSO may affect solubility.
  • Appearance: white to off-white solid.
  • Recommended storage: sealed at -20°C; in solvent store at -80°C for long-term (up to 6 months).
  • Reported purity: 98.1%.

Catalog No. 50-002-89315 Supplier Medchemexpress LLC Supplier No. HY16696810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-COOH (MW 5000) is a PEGylated phospholipid used to modify liposome and nanoparticle surfaces to improve solubility, circulation time, and tissue targeting of poorly soluble drugs.

  • PEGylated phospholipid with an average molecular weight of 5,000 Da.
  • Carboxylic acid terminal group enables covalent conjugation and surface functionalization.
  • Enhances colloidal stability and circulation half-life in delivery systems.
  • Soluble in DMSO at approximately 100 mg/mL with sonication; hygroscopic DMSO may affect solubility.
  • Appearance: white to off-white solid.
  • Recommended storage: sealed at -20°C; in solvent store at -80°C for long-term (up to 6 months).
  • Reported purity: 98.1%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.