Learn More
Creative Pegworks DSPE-PEG-COOH, MW 2K 50MG
SDP
Catalog No. NC1962494
Encompass
DSPE-PEG-COOH is a linear heterobifunctional PEGylation reagent with a DSPE phospholipid and a carboxylic acid. It is a useful self-assembling reagent to prepare PEGylated liposome or micelle while also providing a COOH group for conjugation with amine-containing molecules.50mg CREATIVE PEGWORKS mfg# PLS-9912
Catalog No. NC1962494 Supplier Creative Pegworks Supplier No. PLS991250MG
DSPE-PEG-COOH is a linear heterobifunctional PEGylation reagent with a DSPE phospholipid and a carboxylic acid. It is a useful self-assembling reagent to prepare PEGylated liposome or micelle while also providing a COOH group for conjugation with amine-containing molecules.50mg CREATIVE PEGWORKS mfg# PLS-9912