Learn More
Medchemexpress LLC Dspe-peg-cooh (mw 5000) | 98.1% | 5000 (Average) | 25 MG
SDP
Catalog No. 5000395003
Encompass_Preferred
DSPE-PEG-COOH (MW 5000) is a PEG lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for use in drug delivery research.
  • Can improve delivery efficiency of poorly soluble drugs
  • Enhances tissue specificity
  • White to off-white solid appearance
  • Available in various quantities for research use
Catalog No. 50-003-95003 Supplier Medchemexpress LLC Supplier No. 25MGHY166968
DSPE-PEG-COOH (MW 5000) is a PEG lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for use in drug delivery research.
  • Can improve delivery efficiency of poorly soluble drugs
  • Enhances tissue specificity
  • White to off-white solid appearance
  • Available in various quantities for research use