Learn More
Medchemexpress LLC DSPE-PEG-DBCO | 2052955-83-4 | 99.9% | 2000 Da (average) | (C2H4O)nC43H86NO9P | 10 MG
SDP
Catalog No. 5000280988
Encompass_Preferred
DSPE-PEG-DBCO is a PEGylated phospholipid reagent bearing a dibenzocyclooctyne (DBCO) functional group for strain-promoted alkyne-azide cycloaddition (SPAAC). With an average molecular weight of about 2000 Da and reported high purity, it is designed to introduce a copper-free click handle into lipid formulations, liposomes, and nanoparticles for conjugation to azide-containing biomolecules.
  • Provides a DBCO functional group for copper-free click chemistry.
  • Enables surface functionalization of liposomes and lipid nanoparticles.
  • Average molecular weight ~2000 Da for pegylation compatibility.
  • Reported high purity supports reliable conjugation reactions.
  • Suitable for bioconjugation to azide-bearing biomolecules.
Catalog No. 50-002-80988 Supplier Medchemexpress LLC Supplier No. HYW44083510MG
DSPE-PEG-DBCO is a PEGylated phospholipid reagent bearing a dibenzocyclooctyne (DBCO) functional group for strain-promoted alkyne-azide cycloaddition (SPAAC). With an average molecular weight of about 2000 Da and reported high purity, it is designed to introduce a copper-free click handle into lipid formulations, liposomes, and nanoparticles for conjugation to azide-containing biomolecules.
  • Provides a DBCO functional group for copper-free click chemistry.
  • Enables surface functionalization of liposomes and lipid nanoparticles.
  • Average molecular weight ~2000 Da for pegylation compatibility.
  • Reported high purity supports reliable conjugation reactions.
  • Suitable for bioconjugation to azide-bearing biomolecules.