Learn More
Medchemexpress LLC DSPE-PEG-DBCO sodium | 95.0% | 5000 (average) | 25 MG
SDP
Catalog No. 5000280990
Encompass_Preferred
DSPE-PEG-DBCO sodium (MW 5000) is a PEGylated phospholipid supplied as the sodium salt with an average PEG molecular weight of 5000. It is provided as a solid (white to yellow) for use in nanoparticle and liposome functionalization where copper-free click chemistry is required to attach azide-bearing ligands for targeting or payload conjugation.
  • Enables copper-free click conjugation via DBCO chemistry.
  • Suitable for surface functionalization of lipid nanoparticles and liposomes.
  • Provides PEG shielding to improve circulation and reduce opsonization.
  • Supplied as a solid with reported purity ~95.0%.
  • Average molecular weight 5000 for extended PEG chains and steric stabilization.
Catalog No. 50-002-80990 Supplier Medchemexpress LLC Supplier No. HYW44089425MG
DSPE-PEG-DBCO sodium (MW 5000) is a PEGylated phospholipid supplied as the sodium salt with an average PEG molecular weight of 5000. It is provided as a solid (white to yellow) for use in nanoparticle and liposome functionalization where copper-free click chemistry is required to attach azide-bearing ligands for targeting or payload conjugation.
  • Enables copper-free click conjugation via DBCO chemistry.
  • Suitable for surface functionalization of lipid nanoparticles and liposomes.
  • Provides PEG shielding to improve circulation and reduce opsonization.
  • Supplied as a solid with reported purity ~95.0%.
  • Average molecular weight 5000 for extended PEG chains and steric stabilization.