Learn More
Medchemexpress LLC DSPE-PEG-DBCO sodium | 95.0% | 5000 Da | C65H104N3O12P | 5 MG
SDP
Catalog No. 5000281462
Encompass_Preferred
DSPE-PEG-DBCO sodium (MW 5000) is a PEGylated phospholipid provided as the sodium salt and designed for surface functionalization of liposomes and nanoparticles and for copper-free bioconjugation using DBCO-azide chemistry. It is supplied as a solid for research applications and is intended for use in drug delivery and bioconjugation studies.
  • Provides DBCO functional group for strain-promoted azide-alkyne cycloaddition (copper-free click chemistry).
  • PEG spacer (≈5000 Da) offers steric stabilization and reduces opsonization.
  • Sodium salt form, solid, suitable for incorporation into lipid formulations.
  • Reported high purity (~95.0%) for research-grade applications.
  • Available in small research quantities for formulation and conjugation studies.
Catalog No. 50-002-81462 Supplier Medchemexpress LLC Supplier No. HYW4408945MG
DSPE-PEG-DBCO sodium (MW 5000) is a PEGylated phospholipid provided as the sodium salt and designed for surface functionalization of liposomes and nanoparticles and for copper-free bioconjugation using DBCO-azide chemistry. It is supplied as a solid for research applications and is intended for use in drug delivery and bioconjugation studies.
  • Provides DBCO functional group for strain-promoted azide-alkyne cycloaddition (copper-free click chemistry).
  • PEG spacer (≈5000 Da) offers steric stabilization and reduces opsonization.
  • Sodium salt form, solid, suitable for incorporation into lipid formulations.
  • Reported high purity (~95.0%) for research-grade applications.
  • Available in small research quantities for formulation and conjugation studies.