Learn More
Medchemexpress LLC DSPE-PEG-DBCO sodium | 99.6% | 3400 (average) | 5 MG
SDP
Catalog No. 5000280502
Encompass_Preferred
DSPE-PEG-DBCO sodium (MW 3400) is a DBCO-functionalized PEGylated phospholipid supplied as a white to off-white solid. It provides a bioorthogonal DBCO handle for strain-promoted azide-alkyne cycloaddition (SPAAC) and an amphiphilic DSPE-PEG backbone suitable for incorporation into liposomes and lipid nanoparticles for surface functionalization and delivery research.
  • Provides DBCO handle for copper-free click conjugation.
  • Enables surface functionalization of liposomes and nanoparticles.
  • Amphiphilic PEG-lipid backbone supports stable lipid formulations.
  • PEG chain (average MW 3400) imparts steric stabilization and solubility.
  • Supplied as a solid suitable for small-scale research use.
  • Available in small packaged quantities for laboratory experiments.
Catalog No. 50-002-80502 Supplier Medchemexpress LLC Supplier No. HYW4408815MG
DSPE-PEG-DBCO sodium (MW 3400) is a DBCO-functionalized PEGylated phospholipid supplied as a white to off-white solid. It provides a bioorthogonal DBCO handle for strain-promoted azide-alkyne cycloaddition (SPAAC) and an amphiphilic DSPE-PEG backbone suitable for incorporation into liposomes and lipid nanoparticles for surface functionalization and delivery research.
  • Provides DBCO handle for copper-free click conjugation.
  • Enables surface functionalization of liposomes and nanoparticles.
  • Amphiphilic PEG-lipid backbone supports stable lipid formulations.
  • PEG chain (average MW 3400) imparts steric stabilization and solubility.
  • Supplied as a solid suitable for small-scale research use.
  • Available in small packaged quantities for laboratory experiments.