Learn More
Medchemexpress LLC DSPE-PEG-endo-BCN (MW 2000) | 93.3% | 2000 | 5 MG
SDP
Catalog No. 5000391564
Encompass_Preferred
DSPE-PEG-endo-BCN (MW 2000) is a PEG lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for use in drug delivery research.
  • Peg lipid
  • Can improve delivery efficiency of poorly soluble drugs
  • Can improve tissue specificity of poorly soluble drugs
  • Suitable for drug delivery research
Catalog No. 50-003-91564 Supplier Medchemexpress LLC Supplier No. HY1668745MG
DSPE-PEG-endo-BCN (MW 2000) is a PEG lipid designed to enhance the delivery efficiency and tissue specificity of poorly soluble drugs. It is suitable for use in drug delivery research.
  • Peg lipid
  • Can improve delivery efficiency of poorly soluble drugs
  • Can improve tissue specificity of poorly soluble drugs
  • Suitable for drug delivery research