Learn More
Medchemexpress LLC DSPE-PEG-FITC | 1000788-53-3 | 10,000 (average) | 50 MG
SDP
Catalog No. 5000280994
Encompass_Preferred
DSPE-PEG-FITC (MW 10000) is a fluorescent lipid-PEG conjugate composed of a DSPE lipid anchor linked to a polyethylene glycol (PEG) chain terminated with fluorescein isothiocyanate (FITC). Supplied as a yellow-orange solid, it is used to introduce fluorescent labeling and PEGylation into liposomes, lipid nanoparticles, and other lipid-based formulations for imaging, tracking, and formulation development.
  • Provides fluorescent labeling for liposomes and lipid nanoparticles.
  • Adds PEGylation to improve circulation and reduce nonspecific interactions.
  • Suitable for in vitro and in vivo imaging and tracking applications.
  • Available in multiple package sizes including 50 mg.
  • Yellow to orange solid; soluble in DMSO with sonication and warming.
  • Batch-specific certificate of analysis available for analytical data.
Catalog No. 50-002-80994 Supplier Medchemexpress LLC Supplier No. HYW440914B50MG
DSPE-PEG-FITC (MW 10000) is a fluorescent lipid-PEG conjugate composed of a DSPE lipid anchor linked to a polyethylene glycol (PEG) chain terminated with fluorescein isothiocyanate (FITC). Supplied as a yellow-orange solid, it is used to introduce fluorescent labeling and PEGylation into liposomes, lipid nanoparticles, and other lipid-based formulations for imaging, tracking, and formulation development.
  • Provides fluorescent labeling for liposomes and lipid nanoparticles.
  • Adds PEGylation to improve circulation and reduce nonspecific interactions.
  • Suitable for in vitro and in vivo imaging and tracking applications.
  • Available in multiple package sizes including 50 mg.
  • Yellow to orange solid; soluble in DMSO with sonication and warming.
  • Batch-specific certificate of analysis available for analytical data.