Learn More
Medchemexpress LLC DSPE-PEG-FITC | 1000788-53-3 | 100 MG
SDP
Catalog No. 5000281134
Encompass_Preferred
DSPE-PEG-FITC (MW 10000) is a fluorescein-labeled DSPE-PEG amphiphilic conjugate (average molecular weight 10,000 Da) that forms micelles in aqueous solution. It is commonly used to prepare fluorescent liposomes and nanoparticles for research in drug delivery and imaging, and is supplied for research use only.
  • Provides fluorescein labeling for visualization in fluorescence assays.
  • Forms micelles and incorporates into liposome bilayers for nanoparticle formulation.
  • Average molecular weight supports steric stabilization and extended circulation.
  • Available in laboratory-friendly pack sizes including 100 mg.
  • Supplied with supporting documents such as COA and SDS for quality control.
Catalog No. 50-002-81134 Supplier Medchemexpress LLC Supplier No. HYW440914B100MG
DSPE-PEG-FITC (MW 10000) is a fluorescein-labeled DSPE-PEG amphiphilic conjugate (average molecular weight 10,000 Da) that forms micelles in aqueous solution. It is commonly used to prepare fluorescent liposomes and nanoparticles for research in drug delivery and imaging, and is supplied for research use only.
  • Provides fluorescein labeling for visualization in fluorescence assays.
  • Forms micelles and incorporates into liposome bilayers for nanoparticle formulation.
  • Average molecular weight supports steric stabilization and extended circulation.
  • Available in laboratory-friendly pack sizes including 100 mg.
  • Supplied with supporting documents such as COA and SDS for quality control.