Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dspe-peg-fitc | 1000788-53-3 | 10000 (average) | 5 MG
SDP

Catalog No. 5000281649
Encompass_Preferred

DSPE-PEG-FITC (MW 10000) is a fluorescein-labeled PEGylated phospholipid conjugate (average MW 10,000) used to prepare fluorescent liposomes, micelles, and nanoparticles for tracking and surface functionalization in vitro and in vivo. It is supplied as a yellow-orange solid and is suitable for incorporation into lipid bilayers or nanoparticle surfaces.

  • Fluorescein-labeled PEGylated phospholipid for fluorescent labeling.
  • Average molecular weight 10,000 for consistent PEG chain length.
  • Forms micelles and integrates into liposome bilayers.
  • Soluble in DMSO; use freshly opened DMSO and heat or sonicate to aid dissolution.
  • Store at -20°C and protect from light; follow solvent storage recommendations for solutions.
  • Offered in small-scale quantities suitable for research use.
  • Documentation available: COA, SDS, and data sheet for batch-specific details.

Catalog No. 50-002-81649 Supplier Medchemexpress LLC Supplier No. HYW4409145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-FITC (MW 10000) is a fluorescein-labeled PEGylated phospholipid conjugate (average MW 10,000) used to prepare fluorescent liposomes, micelles, and nanoparticles for tracking and surface functionalization in vitro and in vivo. It is supplied as a yellow-orange solid and is suitable for incorporation into lipid bilayers or nanoparticle surfaces.

  • Fluorescein-labeled PEGylated phospholipid for fluorescent labeling.
  • Average molecular weight 10,000 for consistent PEG chain length.
  • Forms micelles and integrates into liposome bilayers.
  • Soluble in DMSO; use freshly opened DMSO and heat or sonicate to aid dissolution.
  • Store at -20°C and protect from light; follow solvent storage recommendations for solutions.
  • Offered in small-scale quantities suitable for research use.
  • Documentation available: COA, SDS, and data sheet for batch-specific details.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.