Learn More
Medchemexpress LLC Dspe-peg-fitc | 1000788-53-3 | 25 MG
SDP
Catalog No. 5000280993
Encompass_Preferred
DSPE-PEG-FITC (MW 10,000) is a fluorescein-labeled PEGylated phospholipid designed for incorporation into lipid bilayers and nanoparticle surfaces to enable fluorescence imaging and tracking. The amphiphilic conjugate self-assembles in aqueous environments and is commonly used in liposome and micelle formulation for research applications.
  • Provides fluorescent labeling for liposomes and nanoparticles.
  • Amphiphilic structure enables incorporation into bilayers and micelles.
  • Average PEG molecular weight of approximately 10,000 Da.
  • Facilitates visualization and tracking in imaging assays.
  • Supplied as a solid that is light sensitive; store protected from light.
Catalog No. 50-002-80993 Supplier Medchemexpress LLC Supplier No. HYW440914B25MG
DSPE-PEG-FITC (MW 10,000) is a fluorescein-labeled PEGylated phospholipid designed for incorporation into lipid bilayers and nanoparticle surfaces to enable fluorescence imaging and tracking. The amphiphilic conjugate self-assembles in aqueous environments and is commonly used in liposome and micelle formulation for research applications.
  • Provides fluorescent labeling for liposomes and nanoparticles.
  • Amphiphilic structure enables incorporation into bilayers and micelles.
  • Average PEG molecular weight of approximately 10,000 Da.
  • Facilitates visualization and tracking in imaging assays.
  • Supplied as a solid that is light sensitive; store protected from light.