Learn More
Medchemexpress LLC DSPE-PEG-FITC | 1000788-53-3 | 90.3% | 3,400 g/mol (average) | 25 MG
SDP
Catalog No. 5000281192
Encompass_Preferred
DSPE-PEG-FITC (MW 3400) is a fluorescein-labeled PEGylated phospholipid designed to introduce a fluorescent tag into liposomes and lipid nanoparticles for imaging, tracking, and formulation studies.
  • Fluorescently labels lipid bilayers for imaging and tracking.
  • Contains an average PEG molecular weight of 3,400 g/mol.
  • Suitable for incorporation into liposomes and lipid nanoparticles.
  • Provides visible-range fluorescence via FITC for microscopy and flow cytometry.
  • Typical purity ~90.3% as supplied.
  • Available in small laboratory quantities for formulation work.
Catalog No. 50-002-81192 Supplier Medchemexpress LLC Supplier No. HYW44091625MG
DSPE-PEG-FITC (MW 3400) is a fluorescein-labeled PEGylated phospholipid designed to introduce a fluorescent tag into liposomes and lipid nanoparticles for imaging, tracking, and formulation studies.
  • Fluorescently labels lipid bilayers for imaging and tracking.
  • Contains an average PEG molecular weight of 3,400 g/mol.
  • Suitable for incorporation into liposomes and lipid nanoparticles.
  • Provides visible-range fluorescence via FITC for microscopy and flow cytometry.
  • Typical purity ~90.3% as supplied.
  • Available in small laboratory quantities for formulation work.