Learn More
Medchemexpress LLC Dspe-peg-fitc | 1000788-53-3 | 95.0% | 10 MG
SDP
Catalog No. 5000281131
Encompass_Preferred
DSPE-PEG-FITC (MW 1000) is a fluorescein-labeled PEGylated phospholipid supplied as a powder. It functions as a fluorescent amphiphile that incorporates into lipid bilayers or forms micelles for labeling liposomes, nanoparticles, and membranes in fluorescence microscopy, flow cytometry, and tracking applications. Manufacturer documentation provides purity, storage, and handling details.
Catalog No. 50-002-81131 Supplier Medchemexpress LLC Supplier No. HYW44091410MG
DSPE-PEG-FITC (MW 1000) is a fluorescein-labeled PEGylated phospholipid supplied as a powder. It functions as a fluorescent amphiphile that incorporates into lipid bilayers or forms micelles for labeling liposomes, nanoparticles, and membranes in fluorescence microscopy, flow cytometry, and tracking applications. Manufacturer documentation provides purity, storage, and handling details.