Learn More
Medchemexpress LLC DSPE-PEG-FITC | 1000788-53-3 | 95.0% | 100 MG
SDP
Catalog No. 5000281132
Encompass_Preferred
DSPE-PEG-FITC is a fluorescein (FITC)-labeled PEGylated phospholipid (average molecular weight 1000) supplied as a yellow powder for research applications. It is used to introduce a green fluorescent label into liposomes, micelles, and other lipid-based nanoparticles for visualization and tracking in biological studies.
  • Fluorescent tag for green fluorescence detection (FITC).
  • PEGylated phospholipid for stable incorporation into lipid bilayers and micelles.
  • Average molecular weight 1000 to define PEG chain length.
  • Reagent grade purity, 95%.
  • Supplied as a yellow powder with defined cold-storage recommendations.
  • Intended for research use only.
Catalog No. 50-002-81132 Supplier Medchemexpress LLC Supplier No. HYW440914C100MG
DSPE-PEG-FITC is a fluorescein (FITC)-labeled PEGylated phospholipid (average molecular weight 1000) supplied as a yellow powder for research applications. It is used to introduce a green fluorescent label into liposomes, micelles, and other lipid-based nanoparticles for visualization and tracking in biological studies.
  • Fluorescent tag for green fluorescence detection (FITC).
  • PEGylated phospholipid for stable incorporation into lipid bilayers and micelles.
  • Average molecular weight 1000 to define PEG chain length.
  • Reagent grade purity, 95%.
  • Supplied as a yellow powder with defined cold-storage recommendations.
  • Intended for research use only.