Learn More
Medchemexpress LLC Dspe-peg-fitc | 1000788-53-3 | >=95.0% | 100 MG
SDP
Catalog No. 5000280989
Encompass_Preferred
DSPE-PEG-FITC (average MW 10,000) is a fluorescein-labeled PEGylated phospholipid that functions as a fluorescent lipid probe for incorporation into liposomes, micelles, and other lipid-based nanocarriers. Its amphiphilic structure allows integration into lipid bilayers to enable optical labeling, tracking, and imaging of nanoparticles in vitro and in vivo.
  • Fluorescent label FITC for visualization and tracking.
  • Average molecular weight 10,000 Da.
  • Amphiphilic structure integrates into lipid bilayers.
  • Suitable for liposome, micelle, and nanoparticle formulation.
  • Available in multiple package sizes for experimental flexibility.
Catalog No. 50-002-80989 Supplier Medchemexpress LLC Supplier No. HYW440914100MG
DSPE-PEG-FITC (average MW 10,000) is a fluorescein-labeled PEGylated phospholipid that functions as a fluorescent lipid probe for incorporation into liposomes, micelles, and other lipid-based nanocarriers. Its amphiphilic structure allows integration into lipid bilayers to enable optical labeling, tracking, and imaging of nanoparticles in vitro and in vivo.
  • Fluorescent label FITC for visualization and tracking.
  • Average molecular weight 10,000 Da.
  • Amphiphilic structure integrates into lipid bilayers.
  • Suitable for liposome, micelle, and nanoparticle formulation.
  • Available in multiple package sizes for experimental flexibility.