Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-FITC | 95.0% | 600 (average) | 50 MG
SDP

Catalog No. 5000281263
Encompass_Preferred

DSPE-PEG-FITC (MW 600) is a fluorescein-labeled PEGylated lipid used as a fluorescent probe for incorporation into liposomes, lipid nanoparticles, and membrane-labeling applications. It is supplied as a light yellow to yellow solid, has an average molecular weight of ~600, and is intended for laboratory research use.

  • Provides fluorescent labeling via FITC for visualization and tracking.
  • PEGylated lipid anchor enables stable incorporation into membranes and nanoparticles.
  • Average molecular weight approximately 600 for predictable behavior in formulations.
  • Light yellow to yellow solid suitable for standard laboratory handling.
  • Recommended storage at 4°C with protection from light; in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • Purity tested at 95.0% per manufacturer certificate of analysis.

Catalog No. 50-002-81263 Supplier Medchemexpress LLC Supplier No. HYW440914C50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-FITC (MW 600) is a fluorescein-labeled PEGylated lipid used as a fluorescent probe for incorporation into liposomes, lipid nanoparticles, and membrane-labeling applications. It is supplied as a light yellow to yellow solid, has an average molecular weight of ~600, and is intended for laboratory research use.

  • Provides fluorescent labeling via FITC for visualization and tracking.
  • PEGylated lipid anchor enables stable incorporation into membranes and nanoparticles.
  • Average molecular weight approximately 600 for predictable behavior in formulations.
  • Light yellow to yellow solid suitable for standard laboratory handling.
  • Recommended storage at 4°C with protection from light; in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • Purity tested at 95.0% per manufacturer certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.