Learn More
Medchemexpress LLC Dspe-peg-folate | 1236288-25-7 | 98.2% | C65H114N11O16P | 50 MG
SDP
Catalog No. 5000356158
Encompass_Preferred
This PEG derivative, DSPE-PEG-Folate, MW 3350, contains folic acid and targets folate receptors in cancer cells. It can form micelles or lipid bilayers, making it suitable for research in targeted drug delivery systems and capable of prolonging the elimination half-life of drugs.
  • Suitable for targeted drug delivery system research
  • Binds to folate receptors in cancer cells
  • Can form micelles or lipid bilayers
  • Increases elimination half-life of drugs
  • Prolongs blood circulation time
  • Facilitates drug entry into cells via endocytosis pathway
Catalog No. 50-003-56158 Supplier Medchemexpress LLC Supplier No. HY14400950MG
This PEG derivative, DSPE-PEG-Folate, MW 3350, contains folic acid and targets folate receptors in cancer cells. It can form micelles or lipid bilayers, making it suitable for research in targeted drug delivery systems and capable of prolonging the elimination half-life of drugs.
  • Suitable for targeted drug delivery system research
  • Binds to folate receptors in cancer cells
  • Can form micelles or lipid bilayers
  • Increases elimination half-life of drugs
  • Prolongs blood circulation time
  • Facilitates drug entry into cells via endocytosis pathway