Learn More
Medchemexpress LLC DSPE-PEG-Folate | 1236288-25-7 | 98.22% | C65H114N11O16P | 25 MG
SDP
Catalog No. 5000356529
Encompass_Preferred
DSPE-PEG-Folate is a PEG derivative containing folic acid with a targeting effect that binds to folate receptors in cancer cells. It forms micelles/lipid bilayers and can be used for targeted drug delivery system research. Nanoparticles constructed with DSPE-PEG-Folate and anticancer drugs primarily enter cells through the endocytosis pathway mediated by FA receptors. These nanoparticles mainly accumulate in the cytoplasm, and the released drugs are distributed in the cytoplasm and nucleus, effectively delivering drugs to the nucleus. It can also increase the elimination half-life of the drug and prolong blood circulation time.
  • Contains folic acid
  • Binds to folate receptors in cancer cells
  • Forms micelles and lipid bilayers
  • Supports targeted drug delivery system research
  • Enables endocytosis pathway entry for nanoparticles
  • Increases drug elimination half-life
  • Prolongs blood circulation time
Catalog No. 50-003-56529 Supplier Medchemexpress LLC Supplier No. 25MGHY144009
DSPE-PEG-Folate is a PEG derivative containing folic acid with a targeting effect that binds to folate receptors in cancer cells. It forms micelles/lipid bilayers and can be used for targeted drug delivery system research. Nanoparticles constructed with DSPE-PEG-Folate and anticancer drugs primarily enter cells through the endocytosis pathway mediated by FA receptors. These nanoparticles mainly accumulate in the cytoplasm, and the released drugs are distributed in the cytoplasm and nucleus, effectively delivering drugs to the nucleus. It can also increase the elimination half-life of the drug and prolong blood circulation time.
  • Contains folic acid
  • Binds to folate receptors in cancer cells
  • Forms micelles and lipid bilayers
  • Supports targeted drug delivery system research
  • Enables endocytosis pathway entry for nanoparticles
  • Increases drug elimination half-life
  • Prolongs blood circulation time