Learn More
Medchemexpress LLC DSPE-PEG-Folate | 1236288-25-7 | MFCD07107414 | 95.7% | 5000 | 5 MG
SDP
Catalog No. 5000281135
Encompass_Preferred
DSPE-PEG-Folate (MW 5000) is a PEGylated phospholipid in which DSPE is linked to a polyethylene glycol chain terminated with folic acid. It is designed to incorporate into liposomes or nanoparticle surfaces to present folate ligands for receptor-mediated targeting and uptake, and is used in targeted drug delivery, imaging, and nanoparticle functionalization in research settings.
  • Facilitates folate-receptor mediated targeting.
  • Integrates into lipid bilayers and can form micelles.
  • Average molecular weight of 5000 for extended circulation properties.
  • High purity (≈95.7%) suitable for research applications.
  • Stable as a powder when stored at -20°C for long-term storage.
Catalog No. 50-002-81135 Supplier Medchemexpress LLC Supplier No. HYW4408905MG
DSPE-PEG-Folate (MW 5000) is a PEGylated phospholipid in which DSPE is linked to a polyethylene glycol chain terminated with folic acid. It is designed to incorporate into liposomes or nanoparticle surfaces to present folate ligands for receptor-mediated targeting and uptake, and is used in targeted drug delivery, imaging, and nanoparticle functionalization in research settings.
  • Facilitates folate-receptor mediated targeting.
  • Integrates into lipid bilayers and can form micelles.
  • Average molecular weight of 5000 for extended circulation properties.
  • High purity (≈95.7%) suitable for research applications.
  • Stable as a powder when stored at -20°C for long-term storage.