Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-folate | 90.0% | 2000 (Average) | 5 MG
SDP

Catalog No. 5000281137
Encompass_Preferred

DSPE-PEG-folate (average MW 2000) is a PEGylated lipid conjugate in which a DSPE lipid is linked to polyethylene glycol terminating in folate. Supplied as a solid powder for research use, it is used to introduce folate targeting moieties onto liposomes, micelles, and nanoparticle surfaces to enable receptor-mediated delivery to folate receptor-expressing cells. Typical storage: powder at -20°C; in solvent store at -80°C for long-term stability.

  • Provides folate-mediated targeting to folate receptor-expressing cells.
  • Suitable for incorporation into liposomes, micelles, and nanoparticle surfaces.
  • Average molecular weight approximately 2000 Da.
  • Supplied as a solid powder for laboratory research use.
  • Purity approximately 90.0% as provided by the manufacturer.
  • Storage recommendations included for powder and in-solution conditions.

Catalog No. 50-002-81137 Supplier Medchemexpress LLC Supplier No. HYW4408885MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-folate (average MW 2000) is a PEGylated lipid conjugate in which a DSPE lipid is linked to polyethylene glycol terminating in folate. Supplied as a solid powder for research use, it is used to introduce folate targeting moieties onto liposomes, micelles, and nanoparticle surfaces to enable receptor-mediated delivery to folate receptor-expressing cells. Typical storage: powder at -20°C; in solvent store at -80°C for long-term stability.

  • Provides folate-mediated targeting to folate receptor-expressing cells.
  • Suitable for incorporation into liposomes, micelles, and nanoparticle surfaces.
  • Average molecular weight approximately 2000 Da.
  • Supplied as a solid powder for laboratory research use.
  • Purity approximately 90.0% as provided by the manufacturer.
  • Storage recommendations included for powder and in-solution conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.