Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DSPE-PEG-folate | 90.0% | 2000 g/mol (average) | C63H104N9O15P | 10 MG
SDP

Catalog No. 5000281150
Encompass_Preferred

DSPE-PEG-Folate is a PEGylated phospholipid supplied as a powder that presents folic acid as a targeting ligand for folate receptor-mediated delivery. It self-assembles into micelles or lipid bilayers and is intended for use in liposome formulation and targeted drug delivery research.

  • Average molecular weight ~2000 g/mol.
  • Purity 90.0%.
  • Powder; light yellow to yellow.
  • Soluble in DMSO at 50 mg/mL (requires ultrasonic).
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Forms micelles and lipid bilayers for receptor-targeted delivery.

Catalog No. 50-002-81150 Supplier Medchemexpress LLC Supplier No. HYW44088810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DSPE-PEG-Folate is a PEGylated phospholipid supplied as a powder that presents folic acid as a targeting ligand for folate receptor-mediated delivery. It self-assembles into micelles or lipid bilayers and is intended for use in liposome formulation and targeted drug delivery research.

  • Average molecular weight ~2000 g/mol.
  • Purity 90.0%.
  • Powder; light yellow to yellow.
  • Soluble in DMSO at 50 mg/mL (requires ultrasonic).
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Forms micelles and lipid bilayers for receptor-targeted delivery.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.