Learn More
Medchemexpress LLC DSPE-PEG-folate | 95.7% | 5,000 Da (average) | 50 MG
SDP
Catalog No. 5000281130
Encompass_Preferred
DSPE-PEG-Folate is a pegylated lipid conjugate consisting of a DSPE lipid anchor linked to a polyethylene glycol chain terminated with folic acid (average MW 5,000 Da). It is designed to incorporate into lipid bilayers or micelles and provide folate-receptor targeting for nanoparticle and drug-delivery research. Supplied for laboratory formulation and functionalization.
  • Provides folate-mediated targeting to folate receptor-expressing cells.
  • Incorporates into lipid bilayers and micelles for nanoparticle functionalization.
  • Average molecular weight of 5,000 Da for steric stabilization and circulation.
  • High purity suitable for research applications.
  • Available in small research pack sizes for formulation trials.
Catalog No. 50-002-81130 Supplier Medchemexpress LLC Supplier No. HYW44089050MG
DSPE-PEG-Folate is a pegylated lipid conjugate consisting of a DSPE lipid anchor linked to a polyethylene glycol chain terminated with folic acid (average MW 5,000 Da). It is designed to incorporate into lipid bilayers or micelles and provide folate-receptor targeting for nanoparticle and drug-delivery research. Supplied for laboratory formulation and functionalization.
  • Provides folate-mediated targeting to folate receptor-expressing cells.
  • Incorporates into lipid bilayers and micelles for nanoparticle functionalization.
  • Average molecular weight of 5,000 Da for steric stabilization and circulation.
  • High purity suitable for research applications.
  • Available in small research pack sizes for formulation trials.