Learn More
Medchemexpress LLC DSPE-PEG-folate | 95.7% | 5000 (Average) | 10 MG
SDP
Catalog No. 5000281324
Encompass_Preferred
DSPE-PEG-Folate, MW 5000 is a pegylated phospholipid composed of distearoyl phosphatidylethanolamine (DSPE) conjugated to polyethylene glycol terminated with folic acid. It is used to impart folate-receptor targeting to liposomes, micelles, and other nanocarriers for targeted drug delivery and receptor-specific binding studies.
  • Enables folate-receptor mediated targeting of nanocarriers.
  • Average PEG molecular weight of 5000 provides an extended hydrophilic spacer.
  • Integrates into lipid bilayers and forms stable micelles.
  • High purity suitable for research applications.
  • Available in small research pack sizes for formulation studies.
Catalog No. 50-002-81324 Supplier Medchemexpress LLC Supplier No. HYW44089010MG
DSPE-PEG-Folate, MW 5000 is a pegylated phospholipid composed of distearoyl phosphatidylethanolamine (DSPE) conjugated to polyethylene glycol terminated with folic acid. It is used to impart folate-receptor targeting to liposomes, micelles, and other nanocarriers for targeted drug delivery and receptor-specific binding studies.
  • Enables folate-receptor mediated targeting of nanocarriers.
  • Average PEG molecular weight of 5000 provides an extended hydrophilic spacer.
  • Integrates into lipid bilayers and forms stable micelles.
  • High purity suitable for research applications.
  • Available in small research pack sizes for formulation studies.